siRNA Details
| Gene Symbol: Olfr543 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| GenBank mRNA | BC147342.1 | - | 1 | 283 | 9056415 | |
| BC147343.1 | - | 1 | 283 | 9056393 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Olfr543| Gene Symbol: Olfr543 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| GenBank mRNA | BC147342.1 | - | 1 | 283 | 9056415 | |
| BC147343.1 | - | 1 | 283 | 9056393 | ||