siRNA Details
Gene Symbol: Wdr31 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001011976.2 | NP_001011976.2 | 10 | 1069 | ||
XM_006238237.2 | XP_006238299.1 | 10 | 1033 | |||
XM_006238239.2 | XP_006238301.1 | 10 | 1095 | |||
GenBank mRNA | BC079286.1 | - | 10 | 1069 | 7134286 |
