siRNA Details
| Gene Symbol: Tardbp | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001011979.2 | NP_001011979.1 | 5 | 784 | ||
| XM_006239382.2 | XP_006239444.1 | 5 | 837 | |||
| XM_006239384.2 | XP_006239446.1 | 5 | 814 | |||
| XM_006239385.2 | XP_006239447.1 | 5 | 815 | |||
| XM_006239386.2 | XP_006239448.1 | 5 | 819 | |||
| XM_008764234.1 | XP_008762456.1 | 5 | 816 | |||
| XM_008764235.1 | XP_008762457.1 | 5 | 814 | |||
| XM_008764236.1 | XP_008762458.1 | 5 | 814 | |||
| GenBank mRNA | BC083752.1 | - | 5 | 773 | 7192591 | |

Tardbp
Genomic Map