siRNA Details
| Gene Symbol: Lamp3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001012015.1 | NP_001012015.1 | 6 | 1742 | ||
| XM_006248595.2 | XP_006248657.1 | 10 | 2131 | |||
| XM_008768886.1 | XP_008767108.1 | 11 | 2431 | |||
| XM_008768887.1 | XP_008767109.1 | 7 | 1739 | |||
| GenBank mRNA | BC083787.1 | - | 6 | 1742 | 7097568 | |

Lamp3
Genomic Map