siRNA Details
| Gene Symbol: Zscan21 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001012021.2 | NP_001012021.1 | 4 | 1066 | ||
| XM_006249007.2 | XP_006249069.1 | 4 | 1195 | |||
| GenBank mRNA | BC079435.1 | - | 4 | 1014 | 7115285 | |
Please enter the information below and press OK to send your cart to Core Services for purchase.

Zscan21| Gene Symbol: Zscan21 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001012021.2 | NP_001012021.1 | 4 | 1066 | ||
| XM_006249007.2 | XP_006249069.1 | 4 | 1195 | |||
| GenBank mRNA | BC079435.1 | - | 4 | 1014 | 7115285 | |