siRNA Details
| Gene Symbol: Epb4.1l5 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001012023.1 | NP_001012023.1 | 3 | 366 | ||
| GenBank mRNA | BC090012.1 | - | 3 | 366 | 7324591 | |
Please enter the information below and press OK to send your cart to Core Services for purchase.

Epb4.1l5| Gene Symbol: Epb4.1l5 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001012023.1 | NP_001012023.1 | 3 | 366 | ||
| GenBank mRNA | BC090012.1 | - | 3 | 366 | 7324591 | |