siRNA Details
| Gene Symbol: Serpinc1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001012027.1 | NP_001012027.1 | 3, 4 | 687 | ||
| XM_008769647.1 | XP_008767869.1 | 3, 4 | 632 | |||
| GenBank mRNA | AY321346.1 | - | 546 | |||
| BC088467.1 | - | 3, 4 | 687 | 7127992 | ||
| FQ209388.1 | - | 3, 4 | 705 | |||
| FQ209496.1 | - | 3, 4 | 648 | |||
| FQ209598.1 | - | 3, 4 | 673 | |||
| FQ210275.1 | - | 3, 4 | 670 | |||
| FQ210716.1 | - | 3, 4 | 706 | |||
| FQ210762.1 | - | 3, 4 | 705 | |||
| FQ211013.1 | - | 3, 4 | 711 | |||
| FQ211135.1 | - | 3, 4 | 705 | |||
| FQ218922.1 | - | 3, 4 | 716 | |||
| FQ218964.1 | - | 3, 4 | 670 | |||
| FQ219160.1 | - | 3, 4 | 706 | |||

Serpinc1
Genomic Map