siRNA Details
| Gene Symbol: Serpinc1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001012027.1 | NP_001012027.1 | 7 | 1478 | ||
| XM_008769647.1 | XP_008767869.1 | 7 | 1423 | |||
| GenBank mRNA | BC088467.1 | - | 7 | 1478 | 7127992 | |
| FQ209388.1 | - | 7 | 1498 | |||
| FQ209496.1 | - | 7 | 1439 | |||
| FQ209598.1 | - | 7 | 1464 | |||
| FQ210275.1 | - | 7 | 1463 | |||
| FQ210762.1 | - | 7 | 1496 | |||
| FQ211013.1 | - | 7 | 1502 | |||
| FQ211135.1 | - | 7 | 1496 | |||
| FQ218285.1 | - | 7 | 1492 | |||
| FQ218922.1 | - | 7 | 1508 | |||
| FQ218964.1 | - | 7 | 1459 | |||
| FQ219233.1 | - | 7 | 1463 | |||

Serpinc1
Genomic Map