siRNA Details
| Gene Symbol: Arhgap24 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001012032.1 | NP_001012032.1 | 6 | 1789 | ||
| XM_006250652.2 | XP_006250714.1 | 7 | 1995 | |||
| XM_006250654.2 | XP_006250716.1 | 7 | 4577 | |||
| XM_008770017.1 | XP_008768239.1 | 7 | 2656 | |||
| XM_008770018.1 | XP_008768240.1 | 6 | 2508 | |||
| GenBank mRNA | AB108670.1 | - | 6 | 1991 | ||
| BC085797.1 | - | 6 | 1789 | 7109913 | ||

Arhgap24
Genomic Map