siRNA Details
| Gene Symbol: Lias | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001012037.1 | NP_001012037.1 | 10 | 1321 | ||
| GenBank mRNA | BC083708.1 | - | 10 | 1321 | 7190554 | |
Please enter the information below and press OK to send your cart to Core Services for purchase.

Lias| Gene Symbol: Lias | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001012037.1 | NP_001012037.1 | 10 | 1321 | ||
| GenBank mRNA | BC083708.1 | - | 10 | 1321 | 7190554 | |