siRNA Details
| Gene Symbol: DENND4C | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_017925.6 | NP_060395.5 | 31, 32 | 5689 | ||
| NR_073201.3 | 31, 32 | 5555 | ||||
| XM_006716804.3 | XP_006716867.1 | 32, 33 | 5804 | |||
| XM_006716805.3 | XP_006716868.1 | 31, 32 | 5739 | |||
| XM_006716807.3 | XP_006716870.1 | 23, 24 | 4329 | |||
| XM_017014917.1 | XP_016870406.1 | 31, 32 | 5597 | |||
| XM_017014918.1 | XP_016870407.1 | 33, 34 | 6004 | |||
| XM_017014919.1 | XP_016870408.1 | 33, 34 | 5971 | |||
| XM_017014920.1 | XP_016870409.1 | 31, 32 | 5763 | |||
| GenBank mRNA | AK000693.1 | - | 13, 14 | 1892 | ||
| AK001046.1 | - | 8, 9 | 1270 | |||
| AK002020.1 | - | 6, 7 | 854 | |||
| AK025705.1 | - | 10, 11 | 2645 | |||
| AK125842.1 | - | 25, 26 | 4551 | |||
| AK127734.1 | - | 20, 21 | 3965 | |||
| AK299842.1 | - | 18, 19 | 3610 | |||
| AK302901.1 | - | 13, 14 | 3021 | |||
| AL834521.1 | - | 10, 11 | 1964 | |||
| BC144484.1 | - | 28, 29 | 5411 | |||
| BC144485.1 | - | 27, 28 | 4787 | |||

DENND4C
Genomic Map