siRNA Details
| Gene Symbol: Irf9 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001012041.1 | NP_001012041.1 | 9 | 1553 | ||
| XM_006251974.2 | XP_006252036.1 | 9 | 1681 | |||
| XM_006251975.1 | XP_006252037.1 | 9 | 1625 | |||
| XM_008770679.1 | XP_008768901.1 | 9 | 1666 | |||
| GenBank mRNA | BC079454.1 | - | 9 | 1553 | 7121327 | |

Irf9
Genomic Map