siRNA Details
| Gene Symbol: Xkr6 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001012042.1 | NP_001012042.1 | 4 | 2121 | ||
| GenBank mRNA | AY534260.1 | - | 4 | 2121 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Xkr6| Gene Symbol: Xkr6 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001012042.1 | NP_001012042.1 | 4 | 2121 | ||
| GenBank mRNA | AY534260.1 | - | 4 | 2121 | ||