siRNA Details
| Gene Symbol: Tdrd3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001012043.1 | NP_001012043.1 | 14 | 2440 | ||
| GenBank mRNA | BC081929.1 | - | 14 | 2440 | 7113366 | |
| FQ226413.1 | - | 4 | 1070 | |||
| FQ232347.1 | - | 4 | 1070 | |||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Tdrd3| Gene Symbol: Tdrd3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001012043.1 | NP_001012043.1 | 14 | 2440 | ||
| GenBank mRNA | BC081929.1 | - | 14 | 2440 | 7113366 | |
| FQ226413.1 | - | 4 | 1070 | |||
| FQ232347.1 | - | 4 | 1070 | |||