siRNA Details
| Gene Symbol: Lcp1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001012044.1 | NP_001012044.1 | 11 | 1283 | ||
| XM_006252318.2 | XP_006252380.1 | 11 | 1458 | |||
| XM_006252319.2 | XP_006252381.1 | 13 | 1657 | |||
| XM_006252320.2 | XP_006252382.1 | 13 | 1705 | |||
| XM_006252322.2 | XP_006252384.1 | 11 | 2864 | |||
| GenBank mRNA | BC083855.1 | - | 11 | 1283 | 7125327 | |

Lcp1
Genomic Map