siRNA Details
| Gene Symbol: GPATCH2L | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_017926.3 | NP_060396.2 | 10 | 1778 | ||
| NM_017972.3 | NP_060442.2 | 9 | 1656 | |||
| NR_110314.1 | 10 | 1771 | ||||
| XM_006720191.2 | XP_006720254.1 | 10 | 1736 | |||
| XM_017021427.1 | XP_016876916.1 | 11 | 1978 | |||
| XM_017021428.1 | XP_016876917.1 | 11 | 1963 | |||
| GenBank mRNA | AK000696.1 | - | 10 | 1715 | ||
| AK000895.1 | - | 9 | 1121 | |||
| AK055384.1 | - | 1 | 1833 | |||
| AL712469.1 | - | 1 | 220 | |||

GPATCH2L
Genomic Map