siRNA Details
| Gene Symbol: Cage1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001012052.1 | NP_001012052.1 | 5 | 1290 | ||
| XM_006253808.2 | XP_006253870.1 | 5 | 1277 | |||
| XM_006253809.2 | XP_006253871.1 | 4 | 968 | |||
| XM_006253810.2 | XP_006253872.1 | 5 | 1307 | |||
| XM_006253811.2 | XP_006253873.1 | 5 | 1391 | |||
| GenBank mRNA | BC081934.1 | - | 5 | 1290 | 7114574 | |
| FQ211932.1 | - | 5 | 1395 | |||

Cage1
Genomic Map