siRNA Details
| Gene Symbol: Zkscan3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001012053.2 | NP_001012053.2 | 6 | 2186 | ||
| XM_006254096.2 | XP_006254158.1 | 7 | 3020 | |||
| XM_006254099.2 | XP_006254161.1 | 6 | 2517 | |||
| XM_006254101.2 | XP_006254163.1 | 7 | 2314 | |||
| XM_008771720.1 | XP_008769942.1 | 6 | 7592 | |||
| XM_008771721.1 | XP_008769943.1 | 6 | 7775 | |||
| XM_008771723.1 | XP_008769945.1 | 7 | 7518 | |||
| XM_008771724.1 | XP_008769946.1 | 7 | 7400 | |||
| XM_008771725.1 | XP_008769947.1 | 7 | 7583 | |||
| XM_008771726.1 | XP_008769948.1 | 7 | 2497 | |||
| GenBank mRNA | BC089847.1 | - | 6 | 2189 | 7373658 | |

Zkscan3
Genomic Map