siRNA Details
| Gene Symbol: Calml3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001012054.1 | NP_001012054.1 | 1 | 994 | ||
| XM_006254208.2 | XP_006254270.1 | 2 | 1598 | |||
| GenBank mRNA | BC086350.1 | - | 1 | 994 | 7302843 | |
Please enter the information below and press OK to send your cart to Core Services for purchase.

Calml3| Gene Symbol: Calml3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001012054.1 | NP_001012054.1 | 1 | 994 | ||
| XM_006254208.2 | XP_006254270.1 | 2 | 1598 | |||
| GenBank mRNA | BC086350.1 | - | 1 | 994 | 7302843 | |