siRNA Details
Gene Symbol: Mcoln3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001012059.1 | NP_001012059.1 | 7, 8 | 978 | ||
GenBank mRNA | BC079200.1 | - | 7, 8 | 978 | 7131704 |

Please enter the information below and press OK to send your cart to Core Services for purchase.
Gene Symbol: Mcoln3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001012059.1 | NP_001012059.1 | 7, 8 | 978 | ||
GenBank mRNA | BC079200.1 | - | 7, 8 | 978 | 7131704 |