siRNA Details
| Gene Symbol: Tab2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001012062.1 | NP_001012062.1 | 4 | 1248 | ||
| XM_006227617.2 | XP_006227679.1 | 3 | 1281 | |||
| XM_006227618.2 | XP_006227680.1 | 3 | 1627 | |||
| XM_006227620.2 | XP_006227682.1 | 3 | 1278 | |||
| XM_008758608.1 | XP_008756830.1 | 4 | 1345 | |||
| GenBank mRNA | BC085788.1 | - | 4 | 1248 | 7108689 | |

Tab2
Genomic Map