siRNA Details
| Gene Symbol: Tab2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001012062.1 | NP_001012062.1 | 6 | 2058 | ||
| GenBank mRNA | BC085788.1 | - | 6 | 2058 | 7108689 | |
Please enter the information below and press OK to send your cart to Core Services for purchase.

Tab2| Gene Symbol: Tab2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001012062.1 | NP_001012062.1 | 6 | 2058 | ||
| GenBank mRNA | BC085788.1 | - | 6 | 2058 | 7108689 | |