siRNA Details
| Gene Symbol: Sae1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001012063.1 | NP_001012063.1 | 9 | 1655 | ||
| GenBank mRNA | BC079411.1 | - | 9 | 1655 | 7113315 | |
Please enter the information below and press OK to send your cart to Core Services for purchase.

Sae1| Gene Symbol: Sae1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001012063.1 | NP_001012063.1 | 9 | 1655 | ||
| GenBank mRNA | BC079411.1 | - | 9 | 1655 | 7113315 | |