siRNA Details
| Gene Symbol: Pvrl2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001012064.1 | NP_001012064.1 | 9 | 2113 | ||
| GenBank mRNA | BC090075.1 | - | 9 | 2113 | 7189358 | |
Please enter the information below and press OK to send your cart to Core Services for purchase.

Pvrl2| Gene Symbol: Pvrl2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001012064.1 | NP_001012064.1 | 9 | 2113 | ||
| GenBank mRNA | BC090075.1 | - | 9 | 2113 | 7189358 | |