siRNA Details
| Gene Symbol: Ptgdr2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001012070.1 | NP_001012070.1 | 1 | 99 | ||
| XM_006231040.2 | XP_006231102.1 | 1 | 187 | |||
| XM_008760231.1 | XP_008758453.1 | 2 | 165 | |||
| GenBank mRNA | AY228550.1 | - | 1 | 99 | ||
More Information
Other Species Targeted
Set Membership: |
|

Ptgdr2
Genomic Map