siRNA Details
| Gene Symbol: Rrp1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001012073.1 | NP_001012073.1 | 2 | 199 | ||
| GenBank mRNA | BC087152.1 | - | 2 | 199 | 7132167 | |
| Gene Symbol: LOC102553324 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | XM_006249615.2 | XP_006249677.1 | 2 | 193 | ||

Rrp1
Genomic Map