siRNA Details
| Gene Symbol: Herc4 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001012074.1 | NP_001012074.1 | 14 | 2028 | ||
| XM_006256134.2 | XP_006256196.1 | 13 | 2010 | |||
| XM_006256135.2 | XP_006256197.1 | 14 | 2041 | |||
| XM_006256136.2 | XP_006256198.1 | 12 | 1861 | |||
| GenBank mRNA | BC087104.1 | - | 14 | 2028 | 7192894 | |

Herc4
Genomic Map