siRNA Details
| Gene Symbol: C17orf80 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001100621.2 | NP_001094091.1 | 3 | 1148 | ||
| NM_001100622.2 | NP_001094092.1 | 3 | 1148 | |||
| NM_001288770.1 | NP_001275699.1 | 3 | 1309 | |||
| NM_001288771.1 | NP_001275700.1 | 3 | 1309 | |||
| NM_017941.5 | NP_060411.2 | 3 | 1148 | |||
| XM_005257487.3 | XP_005257544.1 | 3 | 1141 | |||
| XM_006721966.3 | XP_006722029.1 | 3 | 1148 | |||
| XM_011524961.1 | XP_011523263.1 | 3 | 1148 | |||
| XM_011524962.2 | XP_011523264.1 | 2 | 1046 | |||
| XM_017024806.1 | XP_016880295.1 | 2 | 1074 | |||
| GenBank mRNA | AK000728.1 | - | 3 | 1107 | ||
| AK292838.1 | - | 3 | 1077 | |||
| AK301905.1 | - | 3 | 1309 | |||
| AL136740.1 | - | 3 | 1115 | |||
| AY163812.1 | - | 3 | 1133 | |||
| BC005005.2 | - | 2 | 1053 | 3544728 | ||

C17orf80
Genomic Map