siRNA Details
| Gene Symbol: Snx7 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001012083.1 | NP_001012083.1 | 8 | 1212 | ||
| XM_006233234.2 | XP_006233296.1 | 8 | 2807 | |||
| GenBank mRNA | BC082033.1 | - | 8 | 1212 | 7130739 | |
Please enter the information below and press OK to send your cart to Core Services for purchase.

Snx7| Gene Symbol: Snx7 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001012083.1 | NP_001012083.1 | 8 | 1212 | ||
| XM_006233234.2 | XP_006233296.1 | 8 | 2807 | |||
| GenBank mRNA | BC082033.1 | - | 8 | 1212 | 7130739 | |