siRNA Details
| Gene Symbol: Cytip | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001012086.1 | NP_001012086.1 | 7 | 628 | ||
| XM_006234202.2 | XP_006234264.1 | 8 | 602 | |||
| GenBank mRNA | BC088207.1 | - | 7 | 628 | 7381695 | |
Please enter the information below and press OK to send your cart to Core Services for purchase.

Cytip| Gene Symbol: Cytip | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001012086.1 | NP_001012086.1 | 7 | 628 | ||
| XM_006234202.2 | XP_006234264.1 | 8 | 602 | |||
| GenBank mRNA | BC088207.1 | - | 7 | 628 | 7381695 | |