siRNA Details
| Gene Symbol: SLC35A5 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_017945.3 | NP_060415.1 | 7 | 1765 | ||
| XM_011512948.1 | XP_011511250.1 | 7 | 1615 | |||
| GenBank mRNA | AK000737.1 | - | 7 | 1668 | ||
| AK001992.1 | - | 2 | 555 | |||
| AK172825.1 | - | 2 | 971 | |||
| AK297411.1 | - | 6 | 1391 | |||
| AL117531.1 | - | 2 | 499 | |||
| AY358994.1 | - | 7 | 1674 | |||
| BC010307.1 | - | 7 | 1727 | 3450563 | ||
| BC013046.1 | - | 7 | 1669 | 3875085 | ||

SLC35A5
Genomic Map