siRNA Details
| Gene Symbol: March7 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001012087.1 | NP_001012087.1 | 5 | 1092 | ||
| XM_008761841.1 | XP_008760063.1 | 5 | 1783 | |||
| XM_008761842.1 | XP_008760064.1 | 4 | 915 | |||
| XM_008761843.1 | XP_008760065.1 | 4 | 915 | |||
| XM_008761844.1 | XP_008760066.1 | 5 | 1373 | |||
| GenBank mRNA | BC083842.1 | - | 5 | 1092 | 7114399 | |
| FQ222785.1 | - | 2 | 662 | |||

March7
Genomic Map