siRNA Details
Gene Symbol: Dusp2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001012089.1 | NP_001012089.1 | 3 | 797 | ||
GenBank mRNA | BC088205.1 | - | 3 | 797 | 7380627 |

Please enter the information below and press OK to send your cart to Core Services for purchase.
Gene Symbol: Dusp2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001012089.1 | NP_001012089.1 | 3 | 797 | ||
GenBank mRNA | BC088205.1 | - | 3 | 797 | 7380627 |