siRNA Details
| Gene Symbol: Tia1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001012096.1 | NP_001012096.1 | 8 | 746 | ||
| XM_006236828.2 | XP_006236890.1 | 9 | 856 | |||
| XM_006236829.2 | XP_006236891.1 | 9 | 855 | |||
| XM_006236830.2 | XP_006236892.1 | 8 | 823 | |||
| XM_006236831.2 | XP_006236893.1 | 9 | 653 | |||
| GenBank mRNA | BC087064.1 | - | 8 | 746 | 7132038 | |

Tia1
Genomic Map