siRNA Details
| Gene Symbol: Ddost | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001012104.1 | NP_001012104.1 | 4 | 441 | ||
| GenBank mRNA | BC082075.1 | - | 4 | 441 | 7133207 | |
| FQ228905.1 | - | 4 | 473 | |||
| FQ229122.1 | - | 4 | 453 | |||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Ddost| Gene Symbol: Ddost | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001012104.1 | NP_001012104.1 | 4 | 441 | ||
| GenBank mRNA | BC082075.1 | - | 4 | 441 | 7133207 | |
| FQ228905.1 | - | 4 | 473 | |||
| FQ229122.1 | - | 4 | 453 | |||