siRNA Details
| Gene Symbol: Galnt14 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001012109.1 | NP_001012109.1 | 6, 7 | 857 | ||
| XM_006239775.2 | XP_006239837.1 | 6, 7 | 1024 | |||
| GenBank mRNA | BC079128.1 | - | 6, 7 | 857 | 7127183 | |

Galnt14
Genomic Map