siRNA Details
| Gene Symbol: Ptdss1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001012113.1 | NP_001012113.1 | 12, 13 | 1411 | ||
| GenBank mRNA | BC087129.1 | - | 12, 13 | 1411 | 7112273 | |
| FQ225243.1 | - | 5, 6 | 353 | |||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Ptdss1| Gene Symbol: Ptdss1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001012113.1 | NP_001012113.1 | 12, 13 | 1411 | ||
| GenBank mRNA | BC087129.1 | - | 12, 13 | 1411 | 7112273 | |
| FQ225243.1 | - | 5, 6 | 353 | |||