siRNA Details
| Gene Symbol: Cbx6 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001012119.1 | NP_001012119.1 | 5 | 2778 | ||
| XM_006241994.2 | XP_006242056.1 | 5 | 2945 | |||
| GenBank mRNA | BC087122.1 | - | 5 | 2778 | 7108347 | |
Please enter the information below and press OK to send your cart to Core Services for purchase.

Cbx6| Gene Symbol: Cbx6 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001012119.1 | NP_001012119.1 | 5 | 2778 | ||
| XM_006241994.2 | XP_006242056.1 | 5 | 2945 | |||
| GenBank mRNA | BC087122.1 | - | 5 | 2778 | 7108347 | |