siRNA Details
| Gene Symbol: Naga | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001012120.1 | NP_001012120.1 | 3 | 216 | ||
| XM_006242087.2 | XP_006242149.1 | 4 | 430 | |||
| XM_006242088.2 | XP_006242150.1 | 4 | 366 | |||
| GenBank mRNA | BC082084.1 | - | 3 | 216 | 7133636 | |

Naga
Genomic Map