siRNA Details
| Gene Symbol: Slc35g2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001012126.1 | NP_001012126.1 | 2 | 1108 | ||
| XM_006243630.2 | XP_006243692.1 | 2 | 1276 | |||
| GenBank mRNA | BC088758.1 | - | 2 | 1108 | 7315776 | |
Please enter the information below and press OK to send your cart to Core Services for purchase.

Slc35g2| Gene Symbol: Slc35g2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001012126.1 | NP_001012126.1 | 2 | 1108 | ||
| XM_006243630.2 | XP_006243692.1 | 2 | 1276 | |||
| GenBank mRNA | BC088758.1 | - | 2 | 1108 | 7315776 | |