siRNA Details
| Gene Symbol: Mapkapk3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001012127.1 | NP_001012127.1 | 6 | 873 | ||
| XM_006243776.2 | XP_006243838.1 | 6 | 963 | |||
| XM_008766531.1 | XP_008764753.1 | 6 | 972 | |||
| GenBank mRNA | BC081974.1 | - | 6 | 873 | 7126359 | |

Mapkapk3
Genomic Map