siRNA Details
| Gene Symbol: Rab34 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001012140.1 | NP_001012140.1 | 4 | 452 | ||
| GenBank mRNA | BC086400.1 | - | 4 | 452 | 7304190 | |
| FQ212591.1 | - | 5 | 502 | |||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Rab34| Gene Symbol: Rab34 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001012140.1 | NP_001012140.1 | 4 | 452 | ||
| GenBank mRNA | BC086400.1 | - | 4 | 452 | 7304190 | |
| FQ212591.1 | - | 5 | 502 | |||