siRNA Details
| Gene Symbol: Tfg | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001012144.1 | NP_001012144.1 | 8 | 1128 | ||
| XM_006248236.2 | XP_006248298.1 | 8 | 1242 | |||
| XM_006248237.2 | XP_006248299.1 | 8 | 1197 | |||
| GenBank mRNA | AB218900.1 | - | 8 | 910 | ||
| BC078947.1 | - | 8 | 1128 | 7109719 | ||

Tfg
Genomic Map