siRNA Details
| Gene Symbol: Hgd | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001012145.1 | NP_001012145.1 | 4, 5 | 426 | ||
| GenBank mRNA | BC078948.1 | - | 4, 5 | 426 | 7109741 | |
Please enter the information below and press OK to send your cart to Core Services for purchase.

Hgd| Gene Symbol: Hgd | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001012145.1 | NP_001012145.1 | 4, 5 | 426 | ||
| GenBank mRNA | BC078948.1 | - | 4, 5 | 426 | 7109741 | |