siRNA Details
| Gene Symbol: Pxn | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001012147.1 | NP_001012147.1 | 4 | 699 | ||
| GenBank mRNA | BC081984.1 | - | 4 | 699 | 7128801 | |
Please enter the information below and press OK to send your cart to Core Services for purchase.

Pxn| Gene Symbol: Pxn | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001012147.1 | NP_001012147.1 | 4 | 699 | ||
| GenBank mRNA | BC081984.1 | - | 4 | 699 | 7128801 | |