siRNA Details
| Gene Symbol: Uchl5 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001012149.1 | NP_001012149.1 | 12 | 1607 | ||
| GenBank mRNA | BC088841.1 | - | 12 | 1607 | 7125859 | |
Please enter the information below and press OK to send your cart to Core Services for purchase.

Uchl5| Gene Symbol: Uchl5 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001012149.1 | NP_001012149.1 | 12 | 1607 | ||
| GenBank mRNA | BC088841.1 | - | 12 | 1607 | 7125859 | |