siRNA Details
| Gene Symbol: Uchl5 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001012149.1 | NP_001012149.1 | 6, 7 | 641 | ||
| XM_006249960.2 | XP_006250022.1 | 6, 7 | 676 | |||
| XM_006249961.2 | XP_006250023.1 | 6, 7 | 676 | |||
| XM_006249962.2 | XP_006250024.1 | 6, 7 | 677 | |||
| XM_006249963.2 | XP_006250025.1 | 6, 7 | 678 | |||
| GenBank mRNA | BC088841.1 | - | 6, 7 | 641 | 7125859 | |

Uchl5
Genomic Map