siRNA Details
| Gene Symbol: Tbrg4 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001012154.1 | NP_001012154.1 | 2, 3 | 770 | ||
| XM_006251342.2 | XP_006251404.1 | 4, 5 | 952 | |||
| XM_006251343.2 | XP_006251405.1 | 4, 5 | 882 | |||
| GenBank mRNA | BC087073.1 | - | 2, 3 | 770 | 7134456 | |

Tbrg4
Genomic Map