siRNA Details
| Gene Symbol: Tbrg4 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001012154.1 | NP_001012154.1 | 10 | 2036 | ||
| XM_006251342.2 | XP_006251404.1 | 12 | 2218 | |||
| XM_006251343.2 | XP_006251405.1 | 12 | 2148 | |||
| GenBank mRNA | BC087073.1 | - | 10 | 2036 | 7134456 | |
| FQ217643.1 | - | 12 | 2181 | |||
| FQ217828.1 | - | 12 | 2181 | |||

Tbrg4
Genomic Map