siRNA Details
| Gene Symbol: Tm9sf1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001012155.1 | NP_001012155.1 | 4 | 1708 | ||
| XM_006251990.2 | XP_006252052.1 | 5 | 1548 | |||
| XM_006251991.2 | XP_006252053.1 | 5 | 1564 | |||
| XM_008770688.1 | XP_008768910.1 | 4 | 1765 | |||
| GenBank mRNA | BC081891.1 | - | 4 | 1708 | 7109579 | |

Tm9sf1
Genomic Map